missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methylphenyl)ethanol, 97%, Thermo Scientific Chemicals

Product Code. 11357975
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7287-82-3
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00046632
InChI Key SDCBYRLJYGORNK-UHFFFAOYNA-N
Synonym 1-2-methylphenyl ethanol, o-tolyl methylcarbinol, 1-2-methylphenyl ethan-1-ol, methyl m-tolyl carbinol, alpha-2-dimethylbenzyl alcohol, 2-tolylethanol, 1-o-tolylethanol, 1-o-tolyl-ethanol, methyl o-tolyl carbinol, 1-o-methylphenyl ethanol
PubChem CID 110953
SMILES CC(O)C1=CC=CC=C1C

Specifications

Boiling Point 78°C to 80°C (2 mmHg)
Refractive Index 1.5315
Quantity 25 g
Beilstein 2205145
IUPAC Name 1-(2-methylphenyl)ethanol
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 1-(2-Methylphenyl)ethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.