missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-4-nitrophenyl)homopiperazine, 97%, Thermo Scientific Chemicals

49.00 € - 289.44 €

Chemical Identifiers

CAS 392660-27-4
Molecular Formula C11H14FN3O2
Molecular Weight (g/mol) 239.25
InChI Key BSSBZGVNKLRQEL-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-nitrophenyl homopiperazine, 1-2-fluoro-4-nitrophenyl-1,4-diazepane, 1-2-fluoro-4-nitrophenyl-1,4-diazaperhydroepine
PubChem CID 22291169
IUPAC Name 1-(2-fluoro-4-nitrophenyl)-1,4-diazepane
SMILES C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15499938
View Documents
Thermo Scientific Alfa Aesar
H63310.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15479938
View Documents
Thermo Scientific Alfa Aesar
H63310.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15489938
View Documents
Thermo Scientific Alfa Aesar
H63310.06
5 g
289.44 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

392660-27-4
239.25
1-2-fluoro-4-nitrophenyl homopiperazine, 1-2-fluoro-4-nitrophenyl-1,4-diazepane, 1-2-fluoro-4-nitrophenyl-1,4-diazaperhydroepine
1-(2-fluoro-4-nitrophenyl)-1,4-diazepane
C11H14FN3O2
BSSBZGVNKLRQEL-UHFFFAOYSA-N
22291169
C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F

Specifications

392660-27-4
UN3259
1-2-fluoro-4-nitrophenyl homopiperazine, 1-2-fluoro-4-nitrophenyl-1,4-diazepane, 1-2-fluoro-4-nitrophenyl-1,4-diazaperhydroepine
C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])F
239.25
239.24
Solid
C11H14FN3O2
10169275
BSSBZGVNKLRQEL-UHFFFAOYSA-N
1-(2-fluoro-4-nitrophenyl)-1,4-diazepane
22291169
97%
1-(2-Fluoro-4-nitrophenyl)homopiperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.