missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane, Certified AR for Analysis, Fisher Chemical™
68.40 € - 110.00 €
Chemical Identifiers
CAS | 107-06-2 |
---|---|
Molecular Formula | C2H4Cl2 |
Molecular Weight (g/mol) | 98.95 |
MDL Number | 963 |
InChI Key | WSLDOOZREJYCGB-UHFFFAOYSA-N |
IUPAC Name | 1,2-dichloroethane |
SMILES | ClCCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10656062
|
Fisher Chemical
D/1751/15 |
1 L |
68.40 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10354980
|
Fisher Chemical
D/1751/17 |
2.5 L |
110.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
107-06-2 | |
98.95 | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
ClCCCl |
C2H4Cl2 | |
963 | |
1,2-dichloroethane |
Specifications
-35°C | |
Colorless | |
Amber Glass Bottle | |
C2H4Cl2 | |
65mmHg 29 | |
WSLDOOZREJYCGB-UHFFFAOYSA-N | |
1,2-dichloroethane | |
98.96g/mol |
107-06-2 | |
81°C | |
Liquid | |
963 | |
0.8 mPaS at 20°C | |
ClCCCl | |
98.95 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,2-Dichloroethane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 1B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title