missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichlorobenzene, Extra Pure, SLR, Fisher Chemical™
556.00 € - 560.00 €
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10438980
|
Fisher Chemical
D/1600/15 |
1 L | Amber glass bottle |
556.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10191610
|
Fisher Chemical
D/1600/17 |
2.5 L | Amber glass bottle |
556.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10765901
|
Fisher Chemical
D/1600/PB17 |
2.5 L | Plastic Coated Glass bottle |
560.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
95-50-1 | |
C6H4Cl2 | |
535 |
Specifications
95-50-1 | |
179°C | |
535 | |
1.3mbar at 20°C |
-15°C | |
C6H4Cl2 | |
147g/mol | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,2-Dichlorobenzene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title