Learn More
1,2-Dichlorobenzene, 99+%, for HPLC, Thermo Scientific Chemicals
27.84 € - 102.51 €
Chemical Identifiers
CAS | 95-50-1 |
---|---|
Molecular Formula | C6H4Cl2 |
Molecular Weight (g/mol) | 147 |
InChI Key | RFFLAFLAYFXFSW-UHFFFAOYSA-N |
Synonym | o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben |
PubChem CID | 7239 |
ChEBI | CHEBI:35290 |
IUPAC Name | 1,2-dichlorobenzene |
SMILES | C1=CC=C(C(=C1)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13498588
|
Thermo Scientific Acros
445641000 |
100 mL | Glass bottle |
27.84 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13488588
|
Thermo Scientific Acros
445640010 |
1 L | Glass Bottle |
102.51 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-50-1 | |
147 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
CHEBI:35290 | |
C1=CC=C(C(=C1)Cl)Cl |
C6H4Cl2 | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
7239 | |
1,2-dichlorobenzene |
Specifications
95-50-1 | |
1.3000g/mL | |
67°C | |
C6H4Cl2 | |
1.5500 to 1.5520 | |
1.3 | |
Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene | |
C1=CC=C(C(=C1)Cl)Cl | |
147 | |
CHEBI:35290 | |
99+% | |
1, 2-Dichlorobenzene |
-15.0°C | |
179.0°C to 180.0°C | |
99% min. (GC) | |
C6H4Cl2 | |
15,3065 | |
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
1,2-dichlorobenzene | |
7239 | |
147 | |
HPLC |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.
May cause
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-425-9
RUO – Research Use Only