Learn More
1,2-Dibromoethane-d{4}, 99%(Isotopic), Thermo Scientific Chemicals
90.32 € - 340.00 €
Chemical Identifiers
CAS | 22581-63-1 |
---|---|
Molecular Formula | C2H4Br2 |
Molecular Weight (g/mol) | 191.886 |
MDL Number | MFCD00037499 |
InChI Key | PAAZPARNPHGIKF-LNLMKGTHSA-N |
Synonym | 1,2-dibromoethane-d4, 1,2-dibromo-1,1,2,2-2h4 ethane, dibromo 2 h? ethane, dibromoethane-d4, ethane-d4, 1,2-dibromo, ethylene-d4 dibromide, 2h4-edb, tetradeutero-1,2-dibromoethane, 1,2-dibromoethane-d4, 99 atom % d, 1,2-dibromoethane-d4 99.0 atom % d |
PubChem CID | 123404 |
IUPAC Name | 1,2-dibromo-1,1,2,2-tetradeuterioethane |
SMILES | C(CBr)Br |
Description
It is employed as a mutagenesis research chemical. Dibromoethane was shown to cause DNA single-strand breaks in isolated rat hepatocytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22581-63-1 | |
191.886 | |
PAAZPARNPHGIKF-LNLMKGTHSA-N | |
123404 | |
C(CBr)Br |
C2H4Br2 | |
MFCD00037499 | |
1,2-dibromoethane-d4, 1,2-dibromo-1,1,2,2-2h4 ethane, dibromo 2 h? ethane, dibromoethane-d4, ethane-d4, 1,2-dibromo, ethylene-d4 dibromide, 2h4-edb, tetradeutero-1,2-dibromoethane, 1,2-dibromoethane-d4, 99 atom % d, 1,2-dibromoethane-d4 99.0 atom % d | |
1,2-dibromo-1,1,2,2-tetradeuterioethane |
Specifications
Liquid | |
Slightly Sweetish | |
22581-63-1 | |
132°C | |
MFCD00037499 | |
1,2-dibromoethane-d4, 1,2-dibromo-1,1,2,2-2h4 ethane, dibromo 2 h? ethane, dibromoethane-d4, ethane-d4, 1,2-dibromo, ethylene-d4 dibromide, 2h4-edb, tetradeutero-1,2-dibromoethane, 1,2-dibromoethane-d4, 99 atom % d, 1,2-dibromoethane-d4 99.0 atom % d | |
PAAZPARNPHGIKF-LNLMKGTHSA-N | |
1,2-dibromo-1,1,2,2-tetradeuterioethane | |
123404 | |
99% |
2.20 g/mL | |
1.536 | |
9°C | |
C2H4Br2 | |
UN1605 | |
Insoluble in water. Soluble in chloroform. | |
C(CBr)Br | |
191.886 | |
191.89 | |
1,2-Dibromoethane-d4, Isotopic |
Safety and Handling
GHS H Statement
H301-H311-H331-H350-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335-H350
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE DIBROMIDE
missing translation for 'einecsNumber' : 245-102-8
missing translation for 'tsca' : No
Recommended Storage : Ambient
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.