missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobutane, 98%, Thermo Scientific Chemicals

Product Code. 11404257
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-98-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000157
InChI Key CZWSZZHGSNZRMW-UHFFFAOYNA-N
Synonym butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
PubChem CID 10792
IUPAC Name 1,2-dibromobutane
SMILES CCC(Br)CBr

Specifications

Melting Point -65°C
Density 1.789
Boiling Point 166°C to 167°C
Flash Point >110°C (230°F)
Refractive Index 1.5141
Quantity 250 g
Beilstein 1731380
Merck Index 14,1566
Formula Weight 215.93
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'