missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibenzoylethane, 98+%, Thermo Scientific Chemicals

Product Code. 11352636
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-71-6
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00037818
InChI Key OSWWFLDIIGGSJV-UHFFFAOYSA-N
Synonym 1,2-dibenzoylethane, 1,4-diphenyl-1,4-butanedione, 1,4-butanedione, 1,4-diphenyl, biphenacyl, diphenacyl, 2,2-biacetophenone, acmc-1bd9f, ethane, 1,2-dibenzoyl, succinophenone diphenacyl, cambridge id 5102527
PubChem CID 136322
IUPAC Name 1,4-diphenylbutane-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)CCC(=O)C2=CC=CC=C2

Specifications

Melting Point 144°C to 148°C
Boiling Point 260°C (15 mmHg)
Quantity 1 g
Beilstein 782937
Solubility Information Soluble in acetone. Insoluble in water.
Formula Weight 238.29
Percent Purity ≥98%
Chemical Name or Material 1,2-Dibenzoylethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.