Learn More
1,2-Diaminopropane, 99%, Thermo Scientific Chemicals
25.80 € - 101.00 €
Chemical Identifiers
CAS | 78-90-0 |
---|---|
Molecular Formula | C3H10N2 |
Molecular Weight (g/mol) | 74.13 |
MDL Number | MFCD00008089 |
InChI Key | AOHJOMMDDJHIJH-UHFFFAOYNA-N |
Synonym | 1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine |
PubChem CID | 6567 |
ChEBI | CHEBI:30630 |
IUPAC Name | propane-1,2-diamine |
SMILES | CC(N)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453267
|
Thermo Scientific Alfa Aesar
B24489.AE |
100 mL |
25.80 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15463267
|
Thermo Scientific Alfa Aesar
B24489.AK |
250 mL |
35.90 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473267
|
Thermo Scientific Alfa Aesar
B24489.AP |
500 mL |
58.80 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15483267
|
Thermo Scientific Alfa Aesar
B24489.AU |
1000 mL |
101.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-90-0 | |
74.13 | |
AOHJOMMDDJHIJH-UHFFFAOYNA-N | |
6567 | |
propane-1,2-diamine |
C3H10N2 | |
MFCD00008089 | |
1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine | |
CHEBI:30630 | |
CC(N)CN |
Specifications
78-90-0 | |
0.869 | |
34°C (93°F) | |
C3H10N2 | |
MFCD00008089 | |
605274 | |
14,7852 | |
AOHJOMMDDJHIJH-UHFFFAOYNA-N | |
propane-1,2-diamine | |
6567 | |
74.13 | |
1,2-Diaminopropane |
∼-37°C | |
119°C to 120°C | |
Characteristic | |
1.446 | |
UN2258 | |
Air sensitive; Hygroscopic | |
1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine | |
CC(N)CN | |
74.13 | |
CHEBI:30630 | |
99% |
Safety and Handling
GHS H Statement
H314-H226-H302-H312
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P374-P380-P501c
H226-H302-H311-H314-H317-H334
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: 1,2-PROPYLENEDIAMINE
missing translation for 'einecsNumber' : 201-155-9
missing translation for 'rtecsNumber' : TX6650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only