Learn More
1,2-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals
45.30 € - 168.00 €
Chemical Identifiers
CAS | 931-17-9 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00003861 |
InChI Key | PFURGBBHAOXLIO-UHFFFAOYSA-N |
Synonym | 1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans |
PubChem CID | 13601 |
ChEBI | CHEBI:24567 |
IUPAC Name | cyclohexane-1,2-diol |
SMILES | C1CCC(C(C1)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488497
|
Thermo Scientific Alfa Aesar
A18572.09 |
10 g |
45.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498497
|
Thermo Scientific Alfa Aesar
A18572.18 |
50 g |
168.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2-Cyclohexanediol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Cyclohexanediol is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
931-17-9 | |
116.16 | |
PFURGBBHAOXLIO-UHFFFAOYSA-N | |
13601 | |
cyclohexane-1,2-diol |
C6H12O2 | |
MFCD00003861 | |
1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans | |
CHEBI:24567 | |
C1CCC(C(C1)O)O |
Specifications
931-17-9 | |
118°C to 120°C (10 mmHg) | |
C6H12O2 | |
1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans | |
PFURGBBHAOXLIO-UHFFFAOYSA-N | |
cyclohexane-1,2-diol | |
13601 | |
116.16 | |
1,2-Cyclohexanediol, cis + trans |
72°C to 76°C | |
>110°C (230°F) | |
MFCD00003861 | |
Soluble in water. | |
C1CCC(C(C1)O)O | |
116.16 | |
CHEBI:24567 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 213-229-8
missing translation for 'rtecsNumber' : GV0220000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.