Learn More
1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals
45.83 €
Chemical Identifiers
CAS | 85-42-7 |
---|---|
Molecular Formula | C8H10O3 |
Molecular Weight (g/mol) | 154.17 |
MDL Number | MFCD00064863,MFCD00674195 |
InChI Key | MUTGBJKUEZFXGO-UHFFFAOYNA-N |
Synonym | hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride |
PubChem CID | 85689 |
ChEBI | CHEBI:103210 |
IUPAC Name | 3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
SMILES | O=C1OC(=O)C2CCCCC12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394376
|
Thermo Scientific Alfa Aesar
L20001.22 |
100 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304386
|
Thermo Scientific Alfa Aesar
L20001.36 |
500 g |
45.83 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85-42-7 | |
154.17 | |
MUTGBJKUEZFXGO-UHFFFAOYNA-N | |
85689 | |
3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
C8H10O3 | |
MFCD00064863,MFCD00674195 | |
hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride | |
CHEBI:103210 | |
O=C1OC(=O)C2CCCCC12 |
Specifications
85-42-7 | |
158°C (17 mmHg) | |
C8H10O3 | |
83213 | |
hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride | |
O=C1OC(=O)C2CCCCC12 | |
154.17 | |
CHEBI:103210 | |
97% |
32°C to 34°C | |
275°C (527°F) | |
MFCD00064863,MFCD00674195 | |
Moisture sensitive | |
MUTGBJKUEZFXGO-UHFFFAOYNA-N | |
3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione | |
85689 | |
154.17 | |
1,2-Cyclohexanedicarboxylic anhydride, Cis + Trans |
Safety and Handling
GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
H317-H318-H334
missing translation for 'einecsNumber' : 201-604-9
missing translation for 'rtecsNumber' : NP6895168
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.