missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)-4-nitrobenzene, 98%, Thermo Scientific Chemicals

58.20 € - 174.00 €

Chemical Identifiers

CAS 5339-26-4
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00007386
InChI Key NTURQZFFJDCTMZ-UHFFFAOYSA-N
Synonym 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
PubChem CID 79266
IUPAC Name 1-(2-bromoethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCBr)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11477617
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15677.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11487617
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15677.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).

Solubility
Soluble in water (partly) and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
Specifications

Chemical Identifiers

5339-26-4
230.061
NTURQZFFJDCTMZ-UHFFFAOYSA-N
79266
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
C8H8BrNO2
MFCD00007386
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
1-(2-bromoethyl)-4-nitrobenzene

Specifications

5339-26-4
116°C (0.2 mmHg)
MFCD00007386
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
NTURQZFFJDCTMZ-UHFFFAOYSA-N
1-(2-bromoethyl)-4-nitrobenzene
79266
98%
67°C to 71°C
C8H8BrNO2
1909321
Soluble in water (partly) and methanol.
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
230.061
230.07
1-(2-Bromoethyl)-4-nitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

missing translation for 'einecsNumber' : 226-271-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only