missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-2-bromobenzene, 95%, Thermo Scientific™

15.47 € - 54.07 €

Chemical Identifiers

CAS 18800-28-7
Molecular Formula C8H8Br2O
Molecular Weight (g/mol) 279.96
MDL Number MFCD00634301
InChI Key LOVVQTBEUIBASP-UHFFFAOYSA-N
Synonym 1-bromo-2-2-bromoethoxy benzene, 1-2-bromoethoxy-2-bromobenzene, o-bromo-2-bromoethoxybenzene, benzene, 1-bromo-2-2-bromoethoxy
PubChem CID 2734928
IUPAC Name 1-bromo-2-(2-bromoethoxy)benzene
SMILES BrCCOC1=CC=CC=C1Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10095982
View Documents
Thermo Scientific Acros
379220050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10177542
View Documents
Thermo Scientific Acros
379220250
25 g Glass bottle
54.07 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

18800-28-7
279.96
LOVVQTBEUIBASP-UHFFFAOYSA-N
2734928
BrCCOC1=CC=CC=C1Br
C8H8Br2O
MFCD00634301
1-bromo-2-2-bromoethoxy benzene, 1-2-bromoethoxy-2-bromobenzene, o-bromo-2-bromoethoxybenzene, benzene, 1-bromo-2-2-bromoethoxy
1-bromo-2-(2-bromoethoxy)benzene

Specifications

18800-28-7
100.0
Colorless to Yellow
Authentic
C8H8Br2O
MFCD00634301
LOVVQTBEUIBASP-UHFFFAOYSA-N
1-bromo-2-(2-bromoethoxy)benzene
2734928
95%
1-(2-Bromoethoxy)-2-bromobenzene
94.0
35.0°C to 36.0°C
160.0°C to 162.0°C (16.0 mmHg)
94% min. (GC)
BrC6H4O(CH2)2Br
1-bromo-2-2-bromoethoxy benzene, 1-2-bromoethoxy-2-bromobenzene, o-bromo-2-bromoethoxybenzene, benzene, 1-bromo-2-2-bromoethoxy
BrCCOC1=CC=CC=C1Br
279.96
279.96
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product may darken during storage

Product Identifier
  • 1-(2-Bromoethoxy)-2-bromobenzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only