Learn More
1,2-Bis(trimethylsiloxy)ethane, 98%, Thermo Scientific Chemicals
40.10 € - 124.00 €
Chemical Identifiers
CAS | 7381-30-8 |
---|---|
Molecular Formula | C8H22O2Si2 |
Molecular Weight (g/mol) | 206.43 |
MDL Number | MFCD00009640 |
InChI Key | JGWFUSVYECJQDT-UHFFFAOYSA-N |
Synonym | 1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50 |
PubChem CID | 81858 |
SMILES | C[Si](C)(C)OCCO[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440789
|
Thermo Scientific Alfa Aesar
H63857.09 |
10 g |
40.10 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15450789
|
Thermo Scientific Alfa Aesar
H63857.18 |
50 g |
124.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7381-30-8 | |
206.43 | |
JGWFUSVYECJQDT-UHFFFAOYSA-N | |
81858 |
C8H22O2Si2 | |
MFCD00009640 | |
1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50 | |
C[Si](C)(C)OCCO[Si](C)(C)C |
Specifications
165°C to 171°C | |
C8H22O2Si2 | |
MFCD00009640 | |
1744217 | |
1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50 | |
C[Si](C)(C)OCCO[Si](C)(C)C | |
206.43 | |
206.43 | |
1.403 | |
0.842 |
7381-30-8 | |
(CH3)3SiOCH2CH2OSi(CH3)3 | |
UN1993 | |
51°C (124°F) | |
JGWFUSVYECJQDT-UHFFFAOYSA-N | |
trimethyl(2-trimethylsilyloxyethoxy)silane | |
81858 | |
98% | |
Moisture sensitive | |
1,2-Bis(trimethylsiloxy)ethane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 230-950-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only