missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)-4-methylpiperazine, 97+%, Thermo Scientific Chemicals

Product Code. 15449057
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-98-5
Molecular Formula C7H17N3
Molecular Weight (g/mol) 143.234
MDL Number MFCD03701701
InChI Key GOWUDHPKGOIDIX-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine
PubChem CID 70284
ChEBI CHEBI:84290
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanamine
SMILES CN1CCN(CC1)CCN

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 143.43
Percent Purity ≥97%
Chemical Name or Material 1-(2-Aminoethyl)-4-methylpiperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'