missing translation for 'onlineSavingsMsg'
Learn More

1-(2,5-Dimethylphenyl)piperazine, 96%, Thermo Scientific™

28.10 € - 105.00 €

Chemische Identifikatoren

CAS 1013-25-8
Molecular Formula C12H19N2
Molecular Weight (g/mol) 191.30
MDL Number MFCD00038378
InChI Key YRIFWVMRUFKWLM-UHFFFAOYSA-O
Synonym 1-2,5-dimethylphenyl piperazine, 1-2,5-xylyl piperazine, piperazine, 1-2,5-dimethylphenyl, 2,5-dimethylphenyl piperazine, acmc-1bpa9, 2,5-dimethylphenylpiperazine, n-2,5-dimethylphenyl piperazine, 1-2,5-dimethylphenyl-piperazine, 4-2,5-dimethyl-phenyl piperazine, 1-2,5-dimethyl-phenyl-piperazine
PubChem CID 70542
IUPAC Name 1-(2,5-dimethylphenyl)piperazine
SMILES CC1=CC=C(C)C(=C1)N1CC[NH2+]CC1
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11337585
Unterlagen ansehen Aktionsdetails
Thermo Scientific Alfa Aesar
L05744.06
5 g
28.10 €
5g
Please to purchase this item. Need a web account? Register with us today!
11347585
Unterlagen ansehen Aktionsdetails
Thermo Scientific Alfa Aesar
L05744.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschreibung

Beschreibung

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spezifikation

Chemische Identifikatoren

1013-25-8
191.30
YRIFWVMRUFKWLM-UHFFFAOYSA-O
70542
CC1=CC=C(C)C(=C1)N1CC[NH2+]CC1
C12H19N2
MFCD00038378
1-2,5-dimethylphenyl piperazine, 1-2,5-xylyl piperazine, piperazine, 1-2,5-dimethylphenyl, 2,5-dimethylphenyl piperazine, acmc-1bpa9, 2,5-dimethylphenylpiperazine, n-2,5-dimethylphenyl piperazine, 1-2,5-dimethylphenyl-piperazine, 4-2,5-dimethyl-phenyl piperazine, 1-2,5-dimethyl-phenyl-piperazine
1-(2,5-dimethylphenyl)piperazine

Spezifikation

42°C to 47°C
154°C (10 mmHg)
MFCD00038378
611229
YRIFWVMRUFKWLM-UHFFFAOYSA-O
1-(2,5-dimethylphenyl)piperazine
70542
96%
1013-25-8
C12H19N2
UN3259
1-2,5-dimethylphenyl piperazine, 1-2,5-xylyl piperazine, piperazine, 1-2,5-dimethylphenyl, 2,5-dimethylphenyl piperazine, acmc-1bpa9, 2,5-dimethylphenylpiperazine, n-2,5-dimethylphenyl piperazine, 1-2,5-dimethylphenyl-piperazine, 4-2,5-dimethyl-phenyl piperazine, 1-2,5-dimethyl-phenyl-piperazine
CC1=CC=C(C)C(=C1)N1CC[NH2+]CC1
191.30
190.28
1-(2,5-Dimethylphenyl)piperazine
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 213-795-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications