missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 17°C |
Color | Colorless |
Density | 1.454g/mL at 25°C |
Boiling Point | 213.5°C |
Quantity | 20 L |
Explosion Limit | 6.6% (150°F) |
Flash Point | 110°C (230°F) |
Ignition Point | 1060°F |
Packaging | Stainless Steel Can |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'