Learn More
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
83.92 € - 342.71 €
Chemical Identifiers
CAS | 4101-68-2 |
---|---|
Molecular Formula | C10H20Br2 |
Molecular Weight (g/mol) | 300.08 |
MDL Number | MFCD00000222 |
InChI Key | GTQHJCOHNAFHRE-UHFFFAOYSA-N |
Synonym | decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo |
PubChem CID | 221483 |
IUPAC Name | 1,10-dibromodecane |
SMILES | BrCCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419493
|
Thermo Scientific Alfa Aesar
A12357.22 |
100 g |
83.92 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429493
|
Thermo Scientific Alfa Aesar
A12357.36 |
500 g |
342.71 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
Chemical Identifiers
4101-68-2 | |
300.08 | |
GTQHJCOHNAFHRE-UHFFFAOYSA-N | |
221483 | |
BrCCCCCCCCCCBr |
C10H20Br2 | |
MFCD00000222 | |
decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo | |
1,10-dibromodecane |
Specifications
4101-68-2 | |
1.34 | |
>110°C (230°F) | |
1.493 | |
506156 | |
Insoluble in water. | |
BrCCCCCCCCCCBr | |
300.08 | |
300.08 | |
1,10-Dibromodecane |
26°C to 30°C | |
157°C to 159°C (14 mmHg) | |
C10H20Br2 | |
MFCD00000222 | |
decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo | |
GTQHJCOHNAFHRE-UHFFFAOYSA-N | |
1,10-dibromodecane | |
221483 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 223-871-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.