missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromo-3,3,3-trifluoroacetone, 95%, Thermo Scientific Chemicals

82.90 € - 306.00 €

Chemical Identifiers

CAS 431-67-4
Molecular Formula C3HBr2F3O
Molecular Weight (g/mol) 269.843
MDL Number MFCD00041362
InChI Key HEPPAPZASXFWTB-UHFFFAOYSA-N
Synonym 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone
PubChem CID 550523
IUPAC Name 3,3-dibromo-1,1,1-trifluoropropan-2-one
SMILES C(C(=O)C(F)(F)F)(Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373006
View Documents
Thermo Scientific Alfa Aesar
L10826.06
5 g
82.90 €
5g
Please to purchase this item. Need a web account? Register with us today!
11383006
View Documents
Thermo Scientific Alfa Aesar
L10826.14
25 g
306.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

Solubility
Soluble in Chloroform. Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Incompatible with strong bases, active metals and strong oxidizing agents.
Specifications

Chemical Identifiers

431-67-4
269.843
HEPPAPZASXFWTB-UHFFFAOYSA-N
550523
C(C(=O)C(F)(F)F)(Br)Br
C3HBr2F3O
MFCD00041362
1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone
3,3-dibromo-1,1,1-trifluoropropan-2-one

Specifications

431-67-4
Pungent
1.4305
UN3265
1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone
HEPPAPZASXFWTB-UHFFFAOYSA-N
3,3-dibromo-1,1,1-trifluoropropan-2-one
550523
95%
111°C to 113°C
C3HBr2F3O
MFCD00041362
636645
Soluble in Chloroform. Not miscible or difficult to mix in water.
C(C(=O)C(F)(F)F)(Br)Br
269.843
269.85
1,1-Dibromo-3,3,3-trifluoroacetone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.