Learn More
1,1,3,3-Tetramethyldisiloxane, 97%, Thermo Scientific Chemicals
42.50 € - 483.00 €
Chemical Identifiers
CAS | 3277-26-7 |
---|---|
Molecular Formula | C4H12OSi2 |
Molecular Weight (g/mol) | 132.309 |
MDL Number | MFCD00008256 |
InChI Key | KWEKXPWNFQBJAY-UHFFFAOYSA-N |
Synonym | 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010 |
PubChem CID | 6327482 |
IUPAC Name | dimethylsilylideneoxonio(dimethyl)silanide |
SMILES | C[Si-](C)[O+]=[Si](C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480108
|
Thermo Scientific Alfa Aesar
B23697.14 |
25 g |
42.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490108
|
Thermo Scientific Alfa Aesar
B23697.22 |
100 g |
110.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444250
|
Thermo Scientific Alfa Aesar
B23697.36 |
500 g |
483.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,1,3,3-Tetramethyldisiloxane is used as a monomer in the production of silicone polymers or silicone resins. It is used as a precursor to prepare other organosilicon compounds. It is also utilized in non-aqueous polymer preparation as well as a laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,3,3-Tetramethyldisiloxane is used as a monomer in the production of silicone polymers or silicone resins. It is used as a precursor to prepare other organosilicon compounds. It is also utilized in non-aqueous polymer preparation as well as a laboratory reagent.
Solubility
It reacts with water to produce dimethylsilanol, dimethylsilanediol and hydrogen gas.
Notes
Moisture sensitive. Keep away from sources of ignition. Store in cool place.
Chemical Identifiers
3277-26-7 | |
132.309 | |
KWEKXPWNFQBJAY-UHFFFAOYSA-N | |
6327482 | |
C[Si-](C)[O+]=[Si](C)C |
C4H12OSi2 | |
MFCD00008256 | |
1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010 | |
dimethylsilylideneoxonio(dimethyl)silanide |
Specifications
70°C to 71°C | |
3277-26-7 | |
MFCD00008256 | |
1733364 | |
1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010 | |
KWEKXPWNFQBJAY-UHFFFAOYSA-N | |
dimethylsilylideneoxonio(dimethyl)silanide | |
6327482 | |
97% | |
1.37 | |
0.76 |
-78°C | |
C4H12OSi2 | |
UN1993 | |
−26°C (−15°F) | |
It reacts with water to produce dimethylsilanol,dimethylsilanediol and hydrogen gas. | |
C[Si-](C)[O+]=[Si](C)C | |
132.309 | |
134.33 | |
Characteristic | |
Moisture sensitive | |
1,1,3,3-Tetramethyldisiloxane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H225
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 221-906-4
missing translation for 'rtecsNumber' : JN1000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only