missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyl-1,3-divinyldisilazane, 96%, Thermo Scientific™

Chemical Identifiers

CAS 7691-02-3
MDL Number MFCD00042846
IUPAC Name [[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,1,3,3-Tetramethyl-1,3-divinyldisilazane is used for silicone rubber, silicone resin, silicone gel and vinyl manufacturing.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

7691-02-3
MFCD00042846
[[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene

Specifications

160°C to 161°C
C8H19NSi2
UN2924
42°C (107°F)
Hydrolyzes in water.
C[Si](C)(C=C)N[Si](C)(C)C=C
185.42
185.42
1.44
0.819
7691-02-3
MFCD00042846
2411661
1,1,3,3-tetramethyl-1,3-divinyldisilazane, 1,3-divinyltetramethyldisilazane, 1,3-divinyl-1,1,3,3-tetramethyldisilazane, silanamine, 1-ethenyl-n-ethenyldimethylsilyl-1,1-dimethyl, bis dimethyl vinyl silyl amine, ccris 2450, bis ethenyldimethylsilyl amine, ethenyl-dimethylsilyl amino-dimethylsilyl ethene, n-dimethylvinylsilyl-1,1-dimethyl-1-vinylsilylamine, dvtmds
WYUIWUCVZCRTRH-UHFFFAOYSA-N
[[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene
82126
96%
Moisture sensitive
1,1,3,3-Tetramethyl-1,3-divinyldisilazane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H226-H302+H332-H311

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 231-701-1

missing translation for 'rtecsNumber' : VV1302250

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.