Learn More
1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 95%, Thermo Scientific Chemicals
48.30 € - 138.00 €
Chemical Identifiers
CAS | 1145-98-8 |
---|---|
Molecular Formula | C16H22Si2 |
Molecular Weight (g/mol) | 270.522 |
MDL Number | MFCD00054786 |
InChI Key | IIOOIYHUTINYQO-UHFFFAOYSA-N |
Synonym | 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane |
PubChem CID | 136916 |
IUPAC Name | [dimethyl(phenyl)silyl]-dimethyl-phenylsilane |
SMILES | C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482110
|
Thermo Scientific Alfa Aesar
H28705.03 |
1 g |
48.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492110
|
Thermo Scientific Alfa Aesar
H28705.06 |
5 g |
138.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
1145-98-8 | |
270.522 | |
IIOOIYHUTINYQO-UHFFFAOYSA-N | |
136916 | |
C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2 |
C16H22Si2 | |
MFCD00054786 | |
1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane | |
[dimethyl(phenyl)silyl]-dimethyl-phenylsilane |
Specifications
111°CC | |
1145-98-8 | |
MFCD00054786 | |
Insoluble in water. | |
C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2 | |
270.522 | |
270.52 | |
1,1,2,2-Tetramethyl-1,2-diphenyldisilane |
34°C to 37°C | |
C16H22Si2 | |
1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane | |
IIOOIYHUTINYQO-UHFFFAOYSA-N | |
[dimethyl(phenyl)silyl]-dimethyl-phenylsilane | |
136916 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.