Learn More
1,1,2,2-Tetrachloroethane, 98+%, Thermo Scientific Chemicals
26.43 € - 53.97 €
Chemical Identifiers
CAS | 79-34-5 |
---|---|
Molecular Formula | C2H2Cl4 |
Molecular Weight (g/mol) | 167.838 |
MDL Number | MFCD00000848 |
InChI Key | QPFMBZIOSGYJDE-UHFFFAOYSA-N |
Synonym | s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan |
PubChem CID | 6591 |
ChEBI | CHEBI:36026 |
IUPAC Name | 1,1,2,2-tetrachloroethane |
SMILES | C(C(Cl)Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321607
|
Thermo Scientific Alfa Aesar
L04489.14 |
25 g |
26.43 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331607
|
Thermo Scientific Alfa Aesar
L04489.22 |
100 g |
27.84 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341607
|
Thermo Scientific Alfa Aesar
L04489.36 |
500 g |
53.97 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.
Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
Chemical Identifiers
79-34-5 | |
167.838 | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
6591 | |
1,1,2,2-tetrachloroethane |
C2H2Cl4 | |
MFCD00000848 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
CHEBI:36026 | |
C(C(Cl)Cl)(Cl)Cl |
Specifications
79-34-5 | |
1.596 | |
C2H2Cl4 | |
1.4935 | |
UN1702 | |
14,9189 | |
Miscible with water,methanol,ethanol,chloroform and acetone. | |
C(C(Cl)Cl)(Cl)Cl | |
167.838 | |
CHEBI:36026 | |
≥98% |
-43°C | |
142°C to 146°C | |
Cl2CHCHCl2 | |
MFCD00000848 | |
969206 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethane | |
6591 | |
167.85 | |
1,1,2,2-Tetrachloroethane |
Safety and Handling
GHS H Statement
H310-H330-H341-H351
Fatal in contact with skin.
Fatal if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
P201-P202-P260-P262-P264b-P270-P271-P281-P284-P302+P350-P304+P340-P308+P313-P310-P361-P363-P501c
H310+H330-H351
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,1,2,2-TETRACHLOROETHANE
missing translation for 'einecsNumber' : 201-197-8
missing translation for 'rtecsNumber' : KI8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.