missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.01M, Reagecon™
BIOPHARMA

Product Code. 11503032 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Packaging:
1L
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured as per the Official Methods of Analysis of AOAC International. Sulfuric Acid, 0.01M, Reagecon™ is a ready-to-use analytical volumetric solution with 0.981g/L H2SO4 in water for use as a laboratory reagent. It is also suitable for use in the food industry.

Sulfuric acid is a highly corrosive strong mineral acid with many uses, including as a laboratory reagent in manual and automated titration. This solution is manufactured as per Official Methods of Analysis of AOAC International. It is also used in cholesterol and glycerol testing in eggs in the food industry.

Specifications

Color Yellow
CAS 7664-93-9, 7732-18-5
Concentration 0.01M
Odor Pungent
Packaging 1L
Normality 0.0832N
Quantity 1 L
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.079g/mol
Chemical Name or Material Sulfuric acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.