missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Countertop and Compac Safety Cabinet

Store smaller quantities near point-of-use with the Justrite Sure-Grip™ EX Countertop and Compac Safety Cabinet.

Specifications

Certifications/Compliance FM, NFPA, OSHA, U (NFPA)
Color Yellow
Door Style Self-close
For Use With (Application) Flammables
Material Steel (CR)
View More Specs

Description

Description

For safely storing smaller quantities of flammable liquids at a workstation or adding to existing storage capacities.

  • One self-closing door
  • One adjustable shelf
  • Color: Yellow

Countertop Cabinets

  • Allow increased access to often-used liquids, without compromising safety
  • Portable size keeps work areas flexible
  • Optional wall hanger assembly available if wall mounting is desired
Compac Cabinets
  • Fit almost anywhere and provide protection even where space is at a premium
  • Holds a full 15 gal. (57L) of flammables

Specifications

Specifications

FM, NFPA, OSHA, U (NFPA)
Self-close
Steel (CR)
1
Yellow
Flammables
1
Epoxy, Polyester
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.