Learn More
Hettich Lab Technology™ Fixed Angle Rotor
Fixed Angle Rotor has a relatively low drag for high-speed centrifugation and oblique angling to speed up sedimentation.
Description
- Used primarily in the higher speed range.
- Relatively low drag.
- Oblique angling of tubes shortens the overall distance that particles must travel, thereby speeding up sedimentation.
Specifications
Fixed Angle Rotor |