missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss. p.a., ACS reagent, absolute alcohol, without additive, A15, ≥99.8%, for license holders only, Honeywell Riedel-de Haën™

Product Code. 15660610
Click to view available options
Quantity:
25 mL
4 x 25 mL
1 L
2.5 L
25 L
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Density 0.789g/mL at 25°C, 0.790g/mL at 20°C
Quantity 1 L
Explosion Limit 19% (60°F)
Flash Point 14°C (57.2°F)
Ignition Point 683°F
Physical Form Absolute Alcohol
Linear Formula CH3CH2OH
MDL Number MFCD00003568
Vapor Pressure 44.6mmHg (20°C)
UN Number UN1170
Beilstein 1718733
Formula Weight 46.07g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.