missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ Harvest Plates

This Opaque 96-well Cell Harvesting Plates has a Glass Fiber membrane used for Cell harvesting.

1740.00 €

Specifications

Certifications/Compliance SBS
Format Standard
Lid Without Lid
Array 8 x 12
Color Opaque
View More Specs

Products 2
Product Code Brand Product Type Pore Size Price Quantity & Availability  
Product Code Brand Product Type Pore Size Price Quantity & Availability  
10186430
View accessory products View Documents
Merck
MAHFB1H60
Multiscreen Harvest-B1; Glass fiber FB 1.0 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10300131
View accessory products View Documents
Merck
MAHFC1H60
Multiscreen Harvest-C1; Glass fiber FC 1.2 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Opaque plates feature choice of Millipore glass fiber B (FB) or glass fiber C (FC) filter media and promote high and uniform flowrates in standard 96-well plate cell harvesters and readers.
  • Made of Barex/TiO2-filled SAN (styrene acrylonitrile) that is resistant to all aqueous solutions including moderate-to-strong acids; fully compatible with standard biosafe scintillants
  • B1 and C1 plates have 100μL well capacity
  • Optimal % open area (for light transmission in coincidence counting) of 26% (in 100μL plates)
  • Filters have an effective filtration area of 0.33cm2 and a pore size of 1.0μm for FB or 1.2μm for FC types
  • Nonsterile; without lid

Order Info

Pack of 60 (in 6 sleeves of 10 plates each)

Compliance

Each lot is quality checked for uniform flowrate and full particulate retention and meets all specifications for SBS Level 3

Specifications

Specifications

SBS
Without Lid
Opaque
96
Cell harvesting
Standard
8 x 12
Barex
Non-sterile
60/Pk.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.