Learn More
Brady™ Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Designed to adhere to highly curved surfaces, wires and cables
Brand:  Brady™ 131587
 View more versions of this product
Product Code. 15173329
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Feature printable zone in white or yellow
  • Transparent, non printable area wraps around wires and cables for laminating printed texts
  • Offers resistance to abrasion, handling, solvents, water, oil and dirt
  • Available in various sizes
Specifications
Specifications
UL Recognised to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL Recognised B-427 labels. Brady sel
Permanent Acrylic
Matte
Label Maker Cartridge
1
Rectangle
25.4 x 101.6 mm
0.102 mm
3/0 AWG to 400 kcmil
1 in.
4 in.
25.40 mm
BMP41/BMP51/BMP53 Labelmaker Labels
Black, White, Clear
Self-laminating Vinyl (B-427)
BMP™51,BMP™53,BMP™41 Printers
1 Ea. (70 labels)
1 x 4 in.
-70°C to +70°C
104.78 mm
101.60 x 25.40 mm
25.4 mm
101.6 mm