missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15648440
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Boiling Point 55°C to 57°C
Quantity 4 L
Flash Point −17°C
Packaging Aluminum Bottle
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Formula Weight 58.08
Percent Purity >99.5%
Grade ACS Reagent
Chemical Name or Material Acetone, ACS Reagent, ≥99.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.