missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linden Fuse for plugs to BS 1362 5 A 1 inch
Brand: Linden H-ECS-FUSE-1362-05-4
Fuse for plugs to BS 1362 5 A 1 inch
Code : 33
Additional Details : Weight : 1.00000kg