missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-hydroxypiperidine, 99%, Thermo Scientific™

Product Code. 11329145
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14813-01-5
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00023734
InChI Key UTTCOAGPVHRUFO-UHFFFAOYNA-N
Synonym 1-benzyl-3-piperidinol, 1-benzyl-3-hydroxypiperidine, n-benzyl-3-hydroxypiperidine, 1-n-benzyl-3-hydroxy-piperidine, 3-piperidinol, 1-phenylmethyl, n-benzyl-3-piperidinol, pubchem7640, pubchem7727, acmc-209xh5, acmc-209xj8
PubChem CID 85773
IUPAC Name 1-benzylpiperidin-3-ol
SMILES OC1CCCN(CC2=CC=CC=C2)C1

Specifications

Density 1.056
Boiling Point 140°C to 142°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 5 g
Beilstein 135964
Sensitivity Hygroscopic
Formula Weight 191.27
Percent Purity 99%
Chemical Name or Material 1-Benzyl-3-hydroxypiperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.