missing translation for 'onlineSavingsMsg'
Learn More

Lauroyl chloride, 98%, Thermo Scientific Chemicals

21.31 € - 146.73 €

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.77
MDL Number MFCD00000740
InChI Key NQGIJDNPUZEBRU-UHFFFAOYSA-N
Synonym lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
PubChem CID 8166
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(Cl)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10308903
View Documents
Thermo Scientific Acros
169030500
50 g Glass bottle
21.31 €
50g
Please to purchase this item. Need a web account? Register with us today!
10419270
View Documents
Thermo Scientific Acros
169032500
250 g Glass bottle
58.69 €
250g
Please to purchase this item. Need a web account? Register with us today!
10615612
View Documents
Thermo Scientific Acros
169030010
1 kg Glass bottle
146.73 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-16-3
218.77
NQGIJDNPUZEBRU-UHFFFAOYSA-N
8166
CCCCCCCCCCCC(Cl)=O
C12H23ClO
MFCD00000740
lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
dodecanoyl chloride

Specifications

112-16-3
0.9200g/mL
140°C
97.5% min. (GC)
CH3(CH2)10COCl
MFCD00000740
0.92
Solubility in water: reacts. Other solubilities: organic solutions
CCCCCCCCCCCC(Cl)=O
218.77
218.77
Liquid
-17.0°C
134.0°C to 137.0°C (11.0 mmHg)
Authentic
C12H23ClO
1.4440 to 1.4460
02, 363
lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
NQGIJDNPUZEBRU-UHFFFAOYSA-N
dodecanoyl chloride
8166
98%
Lauroyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-941-7

Product Identifier
  • Lauroyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.