missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis, Fisher Chemical™

54.35 € - 323.00 €

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10010190
View Documents
Fisher Chemical
J/3720/PB15
1 L HDPE plastic bottle
54.35 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10764181
View Documents
Fisher Chemical
J/3720/17
2.5 L HDPE plastic bottle
79.70 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11452714
View Documents
Fisher Chemical
J/3720/24
10 L Bag in box
323.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis

Specifications

Chemical Identifiers

139-33-3
336.21
ZGTMUACCHSMWAC-UHFFFAOYSA-L
57339238
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
C10H14N2Na2O8
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
CHEBI:64734
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

139-33-3
Liquid
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ZGTMUACCHSMWAC-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
57339238
Colorless
C10H14N2Na2O8
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
336.21
CHEBI:64734
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • EDTA (Diaminoethanetetra-acetic acid, disodium salt solution 0.1 M) (0.2N)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.