Learn More
Di-n-octylamine, 95%, Thermo Scientific Chemicals
33.97 € - 285.42 €
Chemical Identifiers
CAS | 1120-48-5 |
---|---|
Molecular Formula | C16H35N |
Molecular Weight (g/mol) | 241.46 |
MDL Number | MFCD00009557 |
InChI Key | LAWOZCWGWDVVSG-UHFFFAOYSA-N |
Synonym | dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine |
PubChem CID | 3094 |
IUPAC Name | N-octyloctan-1-amine |
SMILES | CCCCCCCCNCCCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10635922
|
Thermo Scientific Acros
117080250 |
25 g | Glass bottle |
33.97 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214820
|
Thermo Scientific Acros
117081000 |
100 g | Glass bottle |
82.41 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10574634
|
Thermo Scientific Acros
117085000 |
500 g | Glass bottle |
285.42 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1120-48-5 | |
241.46 | |
LAWOZCWGWDVVSG-UHFFFAOYSA-N | |
3094 | |
CCCCCCCCNCCCCCCCC |
C16H35N | |
MFCD00009557 | |
dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine | |
N-octyloctan-1-amine |
Specifications
1120-48-5 | |
Colorless to Yellow | |
297°C to 298°C | |
Authentic | |
C16H35N | |
1.4412 to 1.4432 | |
04, 196 | |
dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine | |
CCCCCCCCNCCCCCCCC | |
241.46 | |
241.46 | |
Liquid |
14°C to 15°C | |
0.7900g/mL | |
>110°C | |
96% min. (GC) | |
[CH3(CH2)7]2NH | |
MFCD00009557 | |
0.79 | |
LAWOZCWGWDVVSG-UHFFFAOYSA-N | |
N-octyloctan-1-amine | |
3094 | |
97% | |
Di-n-octylamine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 214-311-6
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.