Learn More
Benzhydrol, 99%, Thermo Scientific Chemicals
32.16 € - 78.09 €
Chemical Identifiers
CAS | 91-01-0 |
---|---|
Molecular Formula | C13H12O |
Molecular Weight (g/mol) | 184.24 |
MDL Number | MFCD00004488 |
InChI Key | QILSFLSDHQAZET-UHFFFAOYSA-N |
Synonym | benzhydrol, diphenylcarbinol, benzohydrol, benzhydryl alcohol, hydroxydiphenylmethane, diphenylmethyl alcohol, diphenyl carbinol, alpha-phenylbenzenemethanol, diphenyl-methanol, benzenemethanol, .alpha.-phenyl |
PubChem CID | 7037 |
IUPAC Name | diphenylmethanol |
SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10060710
|
Thermo Scientific Acros
105391000 |
100 g | Plastic bottle |
32.16 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10417480
|
Thermo Scientific Acros
105395000 |
500 g | Plastic bottle |
78.09 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-01-0 | |
184.24 | |
QILSFLSDHQAZET-UHFFFAOYSA-N | |
7037 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)O |
C13H12O | |
MFCD00004488 | |
benzhydrol, diphenylcarbinol, benzohydrol, benzhydryl alcohol, hydroxydiphenylmethane, diphenylmethyl alcohol, diphenyl carbinol, alpha-phenylbenzenemethanol, diphenyl-methanol, benzenemethanol, .alpha.-phenyl | |
diphenylmethanol |
Specifications
91-01-0 | |
100.0 | |
White to Beige | |
160°C | |
Authentic | |
C13H12O | |
MFCD00004488 | |
15, 1092 | |
Solubility in water: 0.5g/L (20°C) | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)O | |
184.24 | |
184.24 | |
Crystalline Solid |
98.5 | |
65.0°C to 68.0°C | |
297.0°C to 298.0°C | |
0.5% max. (55°C) | |
98.5% min. (GC) | |
(C6H5)2CHOH | |
06, IV, 4648 | |
benzhydrol, diphenylcarbinol, benzohydrol, benzhydryl alcohol, hydroxydiphenylmethane, diphenylmethyl alcohol, diphenyl carbinol, alpha-phenylbenzenemethanol, diphenyl-methanol, benzenemethanol, .alpha.-phenyl | |
QILSFLSDHQAZET-UHFFFAOYSA-N | |
diphenylmethanol | |
7037 | |
99% | |
Benzhydrol, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-033-8
missing translation for 'rtecsNumber' : DC7452000
missing translation for 'tsca' : TSCA
- Benzhydrol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.