Learn More
4'-Isobutylacetophenone, 97%, Thermo Scientific Chemicals
57.08 € - 226.13 €
Chemical Identifiers
CAS | 38861-78-8 |
---|---|
Molecular Formula | C12H16O |
Molecular Weight (g/mol) | 176.26 |
InChI Key | KEAGRYYGYWZVPC-UHFFFAOYSA-N |
Synonym | 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20 |
PubChem CID | 93214 |
IUPAC Name | 1-[4-(2-methylpropyl)phenyl]ethanone |
SMILES | CC(C)CC1=CC=C(C=C1)C(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13439086
|
Thermo Scientific Acros
|
50 g |
57.08 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13498628
|
Thermo Scientific Acros
|
250 g |
226.13 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
38861-78-8 | |
176.26 | |
4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20 | |
1-[4-(2-methylpropyl)phenyl]ethanone |
C12H16O | |
KEAGRYYGYWZVPC-UHFFFAOYSA-N | |
93214 | |
CC(C)CC1=CC=C(C=C1)C(=O)C |
Specifications
38861-78-8 | |
107.0°C (2.0 mmHg) | |
96% min. (GC) | |
C12H16O | |
0.96 | |
KEAGRYYGYWZVPC-UHFFFAOYSA-N | |
1-[4-(2-methylpropyl)phenyl]ethanone | |
93214 | |
97% |
0.9600g/mL | |
54°C | |
Glass Bottle | |
(CH3)2CHCH2C6H4COCH3 | |
4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20 | |
CC(C)CC1=CC=C(C=C1)C(=O)C | |
176.26 | |
176.26 | |
4'-Isobutylacetophenone |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 254-159-8
- 4'-Isobutylacetophenone
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H411-Toxic to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.