Learn More
3-Bromopropionic acid, 97%, Thermo Scientific Chemicals
22.61 € - 180.90 €
Chemical Identifiers
CAS | 590-92-1 |
---|---|
Molecular Formula | C3H5BrO2 |
Molecular Weight (g/mol) | 152.98 |
MDL Number | MFCD00002763 |
InChI Key | DHXNZYCXMFBMHE-UHFFFAOYSA-N |
Synonym | 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r |
PubChem CID | 11553 |
IUPAC Name | 3-bromopropanoic acid |
SMILES | OC(=O)CCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10781921
|
Thermo Scientific Acros
325770100 |
10 g | Glass Bottle |
22.61 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023900
|
Thermo Scientific Acros
325771000 |
100 g | Plastic bottle |
47.94 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657694
|
Thermo Scientific Acros
325775000 |
500 g | Plastic bottle |
180.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-92-1 | |
152.98 | |
DHXNZYCXMFBMHE-UHFFFAOYSA-N | |
11553 | |
OC(=O)CCBr |
C3H5BrO2 | |
MFCD00002763 | |
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r | |
3-bromopropanoic acid |
Specifications
590-92-1 | |
100.0 | |
White to Yellow | |
65°C | |
96% min. (HPLC) | |
BrCH2CH2CO2H | |
02, 256 | |
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r | |
DHXNZYCXMFBMHE-UHFFFAOYSA-N | |
3-bromopropanoic acid | |
11553 | |
97% | |
3-Bromopropionic acid |
96.0 | |
57°C to 63°C | |
140°C to 142°C (45.0 mmHg) | |
Authentic | |
C3H5BrO2 | |
MFCD00002763 | |
15, 1444 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chcl3 and benzene | |
OC(=O)CCBr | |
152.98 | |
152.98 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-694-1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.