missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenol, 98%, Thermo Scientific Chemicals

Product Code. 11302246
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water (24 mg/ml at 25°C).

Notes
Storage temperature: 2 - 4°C. Incompatible with Strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-64-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00020000
InChI Key YPNZJHFXFVLXSE-UHFFFAOYSA-N
Synonym 6-chloro-o-cresol, phenol, 2-chloro-6-methyl, o-cresol, 6-chloro, 2-methyl-6-chlorophenol, 2-chloro-6-methyl-phenol, 6-chloro cresol, 6-chlor-o-kresol, pubchem3669, 6-chloro-2-methylphenol, 2-chloro-6-methyl phenol
PubChem CID 6898
IUPAC Name 2-chloro-6-methylphenol
SMILES CC1=C(C(=CC=C1)Cl)O

Specifications

Density 1.201
Boiling Point 75°C to 76°C (12 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.545
Quantity 25 g
UN Number UN2669
Beilstein 774623
Solubility Information Soluble in water (24mg/ml at 25°C).
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.