missing translation for 'onlineSavingsMsg'
Learn More

1-Bromonaphthalene, 96%, Thermo Scientific Chemicals

50.45 € - 587.93 €

Chemical Identifiers

CAS 90-11-9, 580-13-2
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10234420
View Documents
Thermo Scientific Acros
107151000
100 mL Glass bottle
50.45 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10488050
View Documents
Thermo Scientific Acros
107155000
500 mL Glass bottle
159.80 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11984511
View Documents
Thermo Scientific Acros
107150025
2.5 L Glass bottle
587.93 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90-11-9, 580-13-2
MFCD00003868
naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
1-bromonaphthalene
207.07
DLKQHBOKULLWDQ-UHFFFAOYSA-N
7001
C1=CC=C2C(=C1)C=CC=C2Br

Specifications

90-11-9, 580-13-2
100.0
Authentic
C10H7Br
1.6560 to 1.6590 (20°C, 589nm)
05, 547
naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
C1=CC=C2C(=C1)C=CC=C2Br
207.07
207.07
≥95%
1-Bromonaphthalene, 96%
95.0
Brown-Yellow to Yellow
96%
C10H7Br
MFCD00003868
15, 1436
DLKQHBOKULLWDQ-UHFFFAOYSA-N
1-bromonaphthalene
7001
14.8hPa at 20°C
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

missing translation for 'einecsNumber' : 201-965-2

missing translation for 'rtecsNumber' : QJ1545000

missing translation for 'tsca' : TSCA

Product Identifier
  • 1-Bromonaphthalene

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 1-Bromonaphthalene

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.