Thermo Scientific Water Analysis Solutions
thermo-scientific-water-analysis-solutions thermo-scientific-water-analysis-solutions-m
Bench Meters
orionstar-a214
Thermo Scientific™ Orion Star™ A214 pH/ISE Benchtop Meter
This benchtop meter is an ideal choice for dedicated ISE analysis. Whether you use ISE infrequently or have multiple samples every day, get the information you need quickly and easily from the large, backlit, graphic LCD display. Informative display shows pH/mV/RmV/ISE results and temperature along with electrode status, time, date, sample ID, user ID and calibration points. For advanced users, features such as stability and averaging choices provide additional options.
View the Orion Star A214 meter 
Fisher Scientific Benchtop Meters
Meter Display Type Keypad Number of Channels Meter-powered Stirrer Probe Inputs Datalog USB Output RS232 Output -
Meter
Display Type
TFT color touchscreen with step-by-step text prompts
Keypad
Touchscreen or operate via USB mouse
Number of Channels
1, 2, 3, or 4 depending on model
Meter-powered Stirrer Probe Inputs
2
Datalog
2000 per each parameter, per user login (10)
USB Output
1 (host)
RS232 Output
1
Buy Now
Meter
Display Type
Backlit, Graphic LCD
Keypad
11 keys including 3-soft keys for display specific functions
Number of Channels
1 or 2 depending on model
Meter-powered Stirrer Probe Inputs
1
Datalog
500 with date/time
USB Output
1
RS232 Output
1
Buy Now
Thermo Scientific Benchtop Meters
pH Calibration & Testing with the Thermo Scientific Orion Versa Star
Gayle Gleichauf, Applications Lab Manager, calibrates and tests the pH of local drinking water, using the Thermo Scientific Orion Versa Star benchtop meter. The meter can be set up to include the electrode serial number, the calibration with three buffers, and incremental sample number (pH, millivolts, temperature, method, and more).
Fluoride Calibration & Testing with the Thermo Scientific Orion Versa Star
Gayle Gleichauf, Applications Lab Manager, tests for fluoride in the local tap water, using the Thermo Scientific Orion Versa Star benchtop meter. She demonstrates the various functionalities of the meter, including setting customization, calibration dates, the electrode condition, and more.
Meter Display Type Keypad Number of Channels Meter-powered Stirrer Probe Inputs Datalog USB Output RS232 Output -
Meter
Display Type
Customizable color graphic LCD with step-by-step text prompts
Keypad
Numeric with menu-specific function keys
Number of Channels
1 to 4
Meter-powered Stirrer Probe Inputs
2
Datalog
2000 with date/time
USB Output
2
RS232 Output
1
Buy Now
Meter
Display Type
Graphic LCD with step-by-step text prompts
Keypad
Comprehensive with menu-specific function keys
Number of Channels
1 or 2
Meter-powered Stirrer Probe Inputs
1
Datalog
2000 with date/time
USB Output
1
RS232 Output
1
Buy Now
Meter
Display Type
Graphic LCD with step-by-step text prompts
Keypad
Numeric with menu-specific function keys
Number of Channels
2
Meter-powered Stirrer Probe Inputs
1
Datalog
1000 with date/time
USB Output
1
RS232 Output
1
Buy Now
Meter
Display Type
Fixed segment LCD with basic text prompts
Keypad
Simple with highlighted measure key
Number of Channels
1
Meter-powered Stirrer Probe Inputs
50
Datalog
N/A
USB Output
N/A
RS232 Output
N/A
Buy Now
Portable Meters
Fisher Scientific Portable Meters
Meter Display Type Keypad Number of Channels Datalog USB Output RS232 Output Date/Time, GLP -
Meter
Display Type
Backlit LCD with basic text prompts
Keypad
Simple, 6-key operation
Number of Channels
1
Datalog
200
USB Output
N/A
RS232 Output
N/A
Date/Time, GLP
Yes (AP115 & AP 125)
Buy Now
Meter
Display Type
Custom Dual LCD, (AP74 with backlight)
Keypad
Simple, 8-key operation
Number of Channels
N/A
Datalog
50 (AP72, AP75, AP74), 16 (AP71)
USB Output
N/A
RS232 Output
N/A
Date/Time, GLP
Yes (AP72, AP74, AP75)
Buy Now
Meter
Display Type
Backlit Dual LCD
Keypad
Simple, 8-key operation
Number of Channels
2
Datalog
N/A
USB Output
N/A
RS232 Output
N/A
Date/Time, GLP
N/A
Buy Now
Meter
Display Type
Custom Dual LCD
Keypad
Simple, 8-key operation
Number of Channels
2
Datalog
N/A
USB Output
N/A
RS232 Output
N/A
Date/Time, GLP
N/A
Buy Now
Thermo Scientific Portable Meters
Meter Display Type Keypad Number of Channels Datalog USB Output RS232 Output Date/Time, GLP -
Meter
Display Type
Graphic LCD with step-by-step text prompts
Keypad
Comprehensive with menu-specific function keys
Number of Channels
1 to 3
Datalog
5000 with date/time
USB Output
1
RS232 Output
1
Date/Time, GLP
Yes
Buy Now
Meter
Display Type
Graphic LCD with step-by-step text prompts
Keypad
Comprehensive with menu-specific function keys
Number of Channels
1
Datalog
1000 with date/time
USB Output
1
RS232 Output
1
Date/Time, GLP
Yes
Buy Now
Meter
Display Type
Fixed segment LCD with basic text prompts
Keypad
Simple with highlighted measure key
Number of Channels
1
Datalog
50
USB Output
N/A
RS232 Output
N/A
Date/Time, GLP
N/A
Buy Now
Meter
Display Type
Custom Dual LCD
Keypad
Simple, 8-key operation
Number of Channels
2
Datalog
N/A
USB Output
N/A
RS232 Output
N/A
Date/Time, GLP
N/A
Buy Now
Thermo Scientific Orion Star A123 Dissolved Oxygen Portable Meter
This portable meter combines simplicity with accuracy. A large LCD displays dissolved oxygen readings along with temperature. Icons provide quick updates on battery life and completed calibration status. Simple button layout and onscreen messages help with calibration and setup menu choices.
View the Orion Star A123 meter
Fisher Scientific™ accumet™ AB150
For laboratories that need a basic, accurate, easy-to-use pH/mV meter with exceptionally high accuracy and resolution, the AB150 meter is the perfect choice. Kits include electrodes suitable for use with samples that contain Tris buffer, heavy metals or sulfides.
View all accumet AB150 models
Electrodes & Solutions
electrode-accumet-difference
Fisher Scientific accumet Electrodes
Fisher Scientific™ offers a complete line of accumet™ electrodes for most applications. Choose from ph, ion-selective, metallic, ISE/pH reference, process and biotech electrodes for fast, accurate pH, titration, ORP or ISE measurements.  
Explore Fisher Scientific accumet Electrode
Contact our accumet specialists:
Customer Service: +1 888-358-4706
Email: accumet@fishersci.com
Still can't find what you are looking for or need additional assistance? Call or email our accumet experts. Or visit this link for more information on other accumet products.
Thermo Scientific Orion Electrodes
thermo-scientific-orion-electrodes
The Thermo Scientific™ Orion™ electrode line offers a wide variety of pH, conductivity, ORP and ISE electrodes to deliver accurate, quick and reproducible measurements.
For pH measurement, Ross combination pH electrodes offer unsurpassed accuracy and precision and are compatible with any brand of meter. 
To ensure you choose the electrode that will suit your needs, you will need to consider body materials, body styles and sample requirements. Use this interactive selection tool to help you choose the correct electrode for your application.  
Explore Thermo Scientific Orion Electrodes
Epoxy or Glass?
electrode-epoxy
Epoxy Electrodes
  • Extremely durable, rugged body minimizes breakage
  • Electrode bulb protected by bulb guard
  • Low-maintenance, gel-filled; can be used in higher temperatures than glass electrodes
electrode-glass
Glass Electrodes
  • Compatible with virtually any sample, including solvents
  • Easy to clean
  • Can handle higher temperatures
Water Analysis Solutions
Water Analysis Solutions Demonstration Videos
By use of this code snippet, I agree to the Brightcove Publisher T and C found at https://accounts.brightcove.com/en/terms-and-conditions/. -->
Spectrophotometry
Water Analysis Colorimetry
Orion AquaMate Spectrophotometers
Depending on the needs of your lab, choose the AquaMate 7000 Vis Spectrophotometer for 325 to 1100 nm wavelengths or choose the AquaMate 8000 UV-Vis with full 190 to 1100 nm wavelengths for a greater range of measurement options. Use your current chemistry reagents — no need to change. The AquaMate preprogrammed methods are compatible with many leading reagent manufacturers.
Orion AquaMate Spectrophotometers
Spectrophotometers Optical Design Light Source (Typical lifetime) Detector Wavelength Range Accuracy Repeatability -
Spectrophotometers
Optical Design
Dual beam — internal reference detector
Light Source (Typical lifetime)
1.8nm
Detector
Xenon flash lamp (5 years)
Wavelength
Dual silicon photodiodes
Range
190 to 1100nm
Accuracy
±1.0nm
Repeatability
±0.5nm
Buy Now
Spectrophotometers
Optical Design
Single beam
Light Source (Typical lifetime)
5.0nm
Detector
Tungsten-halogen lamp (1000 hours)
Wavelength
Silicon photodiode
Range
325 to 1100 nm
Accuracy
±1.0nm
Repeatability
±0.5nm
Buy Now
Colorimeters
Thermo Scientific Orion Colorimeters provide portable design with uncompromised performance. Thermo Scientific Orion meters are accurate, dependable and easy to use, with handheld portability, preprogrammed methods, optimum wavelengths, user-friendly displays and easy calibration.
Colorimeters
Colorimeters Incident Light Source Wavelengths Wavelength Accuracy Photometric Accuracy Sample Cell Compatibility Buy Now
Colorimeters
Incident Light Source
6 light emitting diodes (LED)
Wavelengths
430, 530, 560, 580, 610, 660nm
Wavelength Accuracy
±1nm
Photometric Accuracy
±0.005
Sample Cell Compatibility
13mm, 16mm and 24mm round
Buy Now
Colorimeters
Incident Light Source
4 light emitting diodes (LED)
Wavelengths
420, 520, 580, 610nm
Wavelength Accuracy
±2nm
Photometric Accuracy
±0.005A
Sample Cell Compatibility
13mm, 16mm and 24mm round
Buy Now
Turbidity Meters
Thermo Scientific Orion offers turbidity meters. Choose from the Orion AQ4500 Turbidity Meter which offers an all-in-one capability, featuring both white and infrared light sources. The Orion AQ3010 Turbidity Meter, includes a field case, offers faster calibration with step-by-step prompts.
Turbidity Meters
Meters Incident Light Source Measurement Modes Measurement Ranges Resolution Sample Cell Compatibility PowerHeader Warranty Buy Now
Meters
Incident Light Source
Dual Source (White light LED and IR LED)
Measurement Modes
EPA 180.1, ISO nephelometric, ISO absorbance, IR ratiometric, % transmittance, EPA GLI method 2, EBC (European Brewing Chemists), ASBC (American Society of Brewing Chemists)
Measurement Ranges
0 to 4000 NTU EPA 0 to 2000 NTU GLI range 0 to 40 NTU ISO-7027 0 to 150 FAU IR Ratio 0 to 4000 NTU EBC 0 to 24.5 ASBC 0 to 236
Resolution
0.01 (0 to 9.99 NTU), 0.1 (10 to 99.9), 1 (100 to 4000)
Sample Cell Compatibility
24mm round
PowerHeader
4 AA batteries
Warranty
2 years
Buy Now
Meters
Incident Light Source
IR LED
Measurement Modes
ISO nephelometric
Measurement Ranges
0.01 to 1000 NTU
Resolution
0.01 (0.01 to 19.99 NTU), 0.1 (20 to 99.9), 1 (100 to 1000)
Sample Cell Compatibility
25mm round
PowerHeader
4 AAA batteries
Warranty
3 years